siRNA Details
Gene Symbol: A430106A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020770.1 | - | 1 | 170 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724654.1 | - | 1 | 170 | Chr. 10 - 122660721 - 122661278 [+] on Build GRCm38 |
