siRNA Details
Gene Symbol: B230119M05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045454.1 | 2 | 87 | |||
GenBank mRNA | AK020983.1 | - | 2 | 88 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727074.1 | - | 2 | 88 | Chr. X - 134684869 - 134686462 [-] on Build GRCm38 |
