siRNA Details
Gene Symbol: B230119M05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045454.1 | 3 | 397 | |||
GenBank mRNA | AK020983.1 | - | 3 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727074.1 | - | 3 | 398 | Chr. X - 134684869 - 134686462 [-] on Build GRCm38 |
