siRNA Details
Gene Symbol: A930041H05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020950.1 | - | 1 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA217024.1 | - | 1 | 208 | Chr. 13 - 116851476 - 116851934 [-] on Build GRCm38 |
