siRNA Details
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043087.1 | - | 451 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640368.1 | - | 3 | 447 | Chr. 19 - 29488457 - 29521992 [-] on Build GRCm38 |
