siRNA Details
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020826.1 | - | 346 | |||
AK043087.1 | - | 649 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020826.1 | - | 346 | |||
AK043087.1 | - | 649 |