siRNA Details
Gene Symbol: 1110018N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003793.1 | - | 2 | 96 | ||
AK044157.1 | - | 2 | 121 | |||
AY672066.1 | - | 3 | 329 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1110018N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003793.1 | - | 2 | 96 | ||
AK044157.1 | - | 2 | 121 | |||
AY672066.1 | - | 3 | 329 |