siRNA Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036940.1 | - | 1 | 1800 | ||
AK043267.1 | - | 1 | 1646 | |||
AK172512.1 | - | 1 | 1742 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB257305.2 | - | 1 | 515 | Chr. 13 - 83526186 - 83526806 [+] on Build GRCm38 |
