siRNA Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036940.1 | - | 1 | 1972 | ||
AK043267.1 | - | 1 | 1819 | |||
AK172512.1 | - | 1 | 1915 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB257305.2 | - | 1 | 688 | Chr. 13 - 83526186 - 83526806 [+] on Build GRCm38 |
