siRNA Details
Gene Symbol: B830032F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148567.1 | - | 1 | 1061 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA318114.1 | - | 1 | 411 | Chr. 15 - 65988908 - 65989628 [-] on Build GRCm38 |
