siRNA Details
Gene Symbol: 1700121L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045453.1 | 3 | 498 | |||
GenBank mRNA | AK007232.1 | - | 3 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707323.1 | - | 3 | 499 | Chr. X - 104816776 - 104842455 [+] on Build GRCm38 |
