siRNA Details
Gene Symbol: 1700122H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007238.1 | - | 1 | 119 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707333.1 | - | 1 | 119 | Chr. 17 - 6895940 - 6901717 [+] on Build GRCm38 |
