siRNA Details
Gene Symbol: 2610524E12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012139.1 | - | 1 | 156 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV506768.1 | - | 1 | 363 | Chr. 11 - 95997740 - 95998174 [-] on Build GRCm38 |
