siRNA Details
Gene Symbol: Gm31739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076901.1 | - | 1 | 75 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715613.1 | - | 1 | 75 | Chr. 3 - 151988454 - 151990013 [+] on Build GRCm38 |
