siRNA Details
Gene Symbol: 4930520P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036596.1 | 3 | 286 | |||
GenBank mRNA | AK015856.1 | - | 3 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715407.1 | - | 3 | 287 | Chr. 13 - 70232822 - 70248128 [+] on Build GRCm38 |
