siRNA Details
Gene Symbol: Gm40811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154198.1 | - | 1 | 1187 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY763754.1 | - | 1 | 577 | Chr. 10 - 117149290 - 117149979 [+] on Build GRCm38 |
