siRNA Details
Gene Symbol: Gm27008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038087.1 | - | 369 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB631452.1 | - | 4 | 369 | Chr. 3 - 106938960 - 107091802 [-] on Build GRCm38 |
