siRNA Details
Gene Symbol: Gm4719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045952.1 | 1 | 95 | |||
GenBank mRNA | AK131717.1 | - | 1 | 96 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706359.1 | - | 1 | 91 | Chr. 17 - 89379245 - 89384944 [+] on Build GRCm38 |
