siRNA Details
Gene Symbol: 2610307P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045053.1 | 8 | 1430 | |||
GenBank mRNA | AK038887.1 | - | 2 | 203 | ||
BC025054.1 | - | 7 | 1031 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632643.1 | - | 2 | 203 | Chr. 13 - 28460681 - 28521108 [-] on Build GRCm38 |
