siRNA Details
Gene Symbol: A230072E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038885.1 | - | 837 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY750441.1 | - | 4 | 560 | Chr. X - 151295483 - 151344824 [-] on Build GRCm38 |
