siRNA Details
Gene Symbol: 9330188P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145081.1 | - | 293 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734918.1 | - | 2 | 290 | Chr. 14 - 105589551 - 105591677 [+] on Build GRCm38 |
