siRNA Details
Gene Symbol: Gm15417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040403.1 | 1 | 156 | |||
GenBank mRNA | AK161599.1 | - | 1 | 157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU609588.1 | - | 1 | 170 | Chr. 3 - 89391850 - 89398028 [+] on Build GRCm38 |
