siRNA Details
Gene Symbol: Gm32693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039352.1 | - | 3 | 362 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633099.1 | - | 3 | 359 | Chr. 3 - 139778859 - 139780758 [-] on Build GRCm38 |
