siRNA Details
Gene Symbol: Gm5782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079904.1 | - | 2 | 314 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF950010.1 | - | 2 | 315 | Chr. 11 - 43414743 - 43416813 [-] on Build GRCm38 |
