siRNA Details
Gene Symbol: Gm5106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079527.1 | - | 3 | 1063 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB170853.2 | - | 2 | 380 | Chr. 5 - 8773577 - 8789965 [-] on Build GRCm38 |
