siRNA Details
Gene Symbol: LOC102636254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036371.1 | - | 583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB629244.1 | - | 5 | 583 | Chr. 2 - 55411815 - 55422329 [+] on Build GRCm38 |
