siRNA Details
Gene Symbol: 2310041K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009740.1 | - | 1 | 73 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY754692.1 | - | 1 | 84 | Chr. 8 - 71396161 - 71396776 [-] on Build GRCm38 |
