siRNA Details
Gene Symbol: 1700012C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005895.1 | - | 1 | 20 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706034.1 | - | 1 | 20 | Chr. 6 - 30460985 - 30463313 [+] on Build GRCm38 |
