siRNA Details
Gene Symbol: Gm16249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135904.1 | - | 3 | 275 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719831.1 | - | 3 | 275 | Chr. 13 - 54778086 - 54783091 [-] on Build GRCm38 |
