siRNA Details
Gene Symbol: Gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009290.1 | - | 317 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709079.1 | - | 2 | 320 | Chr. 17 - 35122751 - 35126433 [-] on Build GRCm38 |
