siRNA Details
Gene Symbol: Gm15555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146213.1 | - | 3 | 109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY737855.1 | - | 3 | 109 | Chr. 16 - 76000784 - 76034536 [+] on Build GRCm38 |
