siRNA Details
Gene Symbol: Gm15555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146213.1 | - | 4 | 647 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY758174.1 | - | 3 | 591 | Chr. 16 - 76001315 - 76034602 [+] on Build GRCm38 |
