siRNA Details
Gene Symbol: Gm14123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076437.1 | - | 2 | 210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614001.1 | - | 2 | 210 | Chr. 2 - 150606229 - 150608236 [+] on Build GRCm38 |
