siRNA Details
Gene Symbol: 9530077C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020648.1 | - | 306 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722269.1 | - | 1 | 306 | Chr. 4 - 139192232 - 139192680 [-] on Build GRCm38 |
