siRNA Details
Gene Symbol: 9530075M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020644.1 | - | 1 | 53 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722265.1 | - | 1 | 53 | Chr. 17 - 47327115 - 47327632 [+] on Build GRCm38 |
