siRNA Details
Gene Symbol: 1700066B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040465.1 | 5 | 454 | |||
GenBank mRNA | AK006899.1 | - | 3 | 186 | ||
BC048628.1 | - | 5 | 454 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU962972.1 | - | 5 | 457 | Chr. 1 - 39842430 - 39847332 [-] on Build GRCm38 |
