siRNA Details
Gene Symbol: 1700066B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040465.1 | 1, 2 | 29 | |||
GenBank mRNA | BC048628.1 | - | 1, 2 | 29 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU962972.1 | - | 1, 2 | 32 | Chr. 1 - 39842430 - 39847332 [-] on Build GRCm38 |
