siRNA Details
Gene Symbol: 1700003G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040720.1 | 4 | 524 | |||
GenBank mRNA | AK005635.1 | - | 4 | 525 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705830.1 | - | 4 | 520 | Chr. 9 - 45318305 - 45322075 [-] on Build GRCm38 |
