siRNA Details
Gene Symbol: LOC73899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014661.1 | - | 2 | 731 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714157.1 | - | 2 | 731 | Chr. 19 - 53460622 - 53462163 [+] on Build GRCm38 |
