siRNA Details
Gene Symbol: Gm15270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014660.1 | - | 3 | 329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714156.1 | - | 3 | 326 | Chr. 10 - 24592289 - 24595810 [-] on Build GRCm38 |
