siRNA Details
Gene Symbol: 4833408A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014662.1 | - | 1 | 729 | ||
AK079024.1 | - | 1 | 728 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM932530.1 | - | 1 | 613 | Chr. 19 - 3999751 - 4000427 [-] on Build GRCm38 |
