siRNA Details
Gene Symbol: 4833408A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014662.1 | - | 1 | 605 | ||
AK079024.1 | - | 1 | 604 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714158.1 | - | 1 | 603 | Chr. 19 - 3999727 - 4000540 [-] on Build GRCm38 |
