siRNA Details
Gene Symbol: Gm4221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163160.1 | - | 3, 4 | 321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640347.1 | - | 3, 4 | 318 | Chr. 18 - 54990312 - 55076794 [+] on Build GRCm38 |
