siRNA Details
Gene Symbol: 4930504D19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019668.1 | - | 195 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715236.1 | - | 1 | 195 | Chr. 6 - 84535876 - 84536323 [+] on Build GRCm38 |
