siRNA Details
Gene Symbol: 4930505K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019669.1 | - | 1 | 97 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715248.1 | - | 1 | 89 | Chr. 5 - 148777970 - 148783090 [+] on Build GRCm38 |
