siRNA Details
Gene Symbol: 4930503F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019667.1 | - | 11 | |||
AK076784.1 | - | 11 | ||||
BY715229.1 | - | 11 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615264.1 | - | 11 | Chr. 14 - 33808088 - 33811283 [-] on Build GRCm38 |
