siRNA Details
Gene Symbol: 2900052O17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013671.1 | - | 1 | 726 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712650.1 | - | 1 | 726 | Chr. 10 - 32097535 - 32098349 [-] on Build GRCm38 |
