siRNA Details
Gene Symbol: 4930554G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016119.1 | - | 676 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715721.1 | - | 4 | 676 | Chr. 11 - 14072772 - 14179041 [-] on Build GRCm38 |
