siRNA Details
Gene Symbol: D730045A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045390.1 | 1 | 172 | |||
GenBank mRNA | AK021341.1 | - | 1 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730926.1 | - | 1 | 169 | Chr. 18 - 74017910 - 74019478 [+] on Build GRCm38 |
