siRNA Details
Gene Symbol: D730047N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021346.1 | - | 1 | 405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730938.1 | - | 1 | 394 | Chr. 6 - 17578078 - 17578713 [+] on Build GRCm38 |
